Learn More
STEMCELL Technologies G418
Product Code. 17120002
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity Unit Size
17120002 250 mg 250mg
1 options
Product Code. 17120002 Supplier STEMCELL Technologies Supplier No. 03812
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Antibiotic for selecting transfected mammalian cells
  • Antibiotic G418 is an aminoglycoside related to gentamicin and is toxic to bacteria, yeast, higher plants, mammalian cells, protozoans, and helminths
  • G418 binds to 80S ribosomes in prokaryotic and eukaryotic cells and inhibits protein synthesis, resulting in cell death
  • Resistance to the antibiotic is conferred by aminoglycoside-3-phosphotransferase
  • Vectors containing the G418 resistance gene (neo) are commonly used for transfecting eukaryotic cells, with selection of transfected cells achieved by culturing them in the presence of G418
  • Cells may exhibit variable resistance to the antibiotic and can take up to a week to die
  • The G418 antibiotic is suitable for use in cell culture for the selection and maintenance of G418-resistant clones when using ClonaCell™-TCS Medium (Catalog #03814), ClonaCell™-CHO CD Medium (Catalog #03815), ClonaCell™-CHO ACF Medium (Catalog #03816), ClonaCell™­CHO CD Liquid Medium (Catalog #03817), or other cell culture media
Specifications
CAS 108321-42-2
Quantity 250 mg