missing translation for 'onlineSavingsMsg'
Learn More

G3BP2 Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech CL555-16276-100UL

 View more versions of this product

Product Code. 17689896

  • £375.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

G3BP2 Polyclonal antibody apecifically detetects G3BP2 antigen in Human Samples. It is validated for Immunofluorescence.
Specifications

Specifications

G3BP2
Polyclonal
CoraLite 555
BC011731
Rabbit
Antigen affinity purification
Primary
Human
Liquid
Immunofluorescence
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
G3BP2
G3BP2 fusion protein Ag9355
100 μL
9908
Store at -20°C. Avoid exposure to light.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.