missing translation for 'onlineSavingsMsg'
Learn More

G3335, TRC

Product Code. 30399875
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399875 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399875 Supplier Toronto Research Chemicals Supplier No. TRCG10050010MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Tryptophyl-L-glutamic Acid
CAS 36099-95-3
Molecular Formula C16H19N3O5
Quantity 10 mg
Synonym 67: PN: EP2161028 PAGE: 10 claimed protein, 7: PN: JP2014079213 TABLE: 2 claimed sequence, G 3335, L-Tryptophyl-L-glutamic Acid, L-α-Tryptophanyl-L-glutamic Acid, Tryptophylglutamic Acid
InChI Formula InChI=1S/C16H19N3O5/c17-11(7-9-8-18-12-4-2-1-3-10(9)12)15(22)19-13(16(23)24)5-6-14(20)21/h1-4,8,11,13,18H,5-7,17H2,(H,19,22)(H,20,21)(H,23,24)/t11-,13-/m0/s1
SMILES C([C@@H](C(N[C@@H](CCC(O)=O)C(O)=O)=O)N)C=1C=2C(NC1)=CC=CC2
IUPAC Name L-tryptophyl-L-glutamic acid
Molecular Weight (g/mol) 333.34
Formula Weight 333.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.