Learn More
Invitrogen™ G2E3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589808
Description
Immunogen sequence: MNESKPGDSQ NLACVFCRKH DDCPNKYGEK KTKEKWNLTV HYYCLLMSSG IWQRGKEEEG VYGFLIEDIR KEVNRASKLK CCVCKKNGAS IGCVAPRCKR SYHFPCGLQR ECIFQFTGNF ASFCWDHRPV QIITSNNYRE SLPCTICLEF IEPIPSYNIL RSPCCKNAWF HRDCLQVQAI; Positive Samples: U-87MG, LO2, 293T, OVCAR3, Mouse eye, Mouse brain, Mouse liver, Rat testis; Cellular Location: Cytoplasm, Nucleus, nucleolus.
E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Essential in early embryonic development to prevent apoptotic death.
Specifications
| G2E3 | |
| Polyclonal | |
| Unconjugated | |
| G2E3 | |
| 6030408C04Rik; 9030416F18; AW046379; D930034K21Rik; G2/M phase-specific E3 ubiquitin-protein ligase; G2/M phase-specific HECT-type E3 ubiquitin transferase; G2/M-phase specific E3 ubiquitin ligase; G2/M-phase specific E3 ubiquitin protein ligase; G2e3; hypothetical protein LOC508575; KIAA1333; mKIAA1333; PHD finger protein 7B; PHF7B; RGD1310263 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 217558, 299002, 55632 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5RJY2, Q7L622 | |
| G2E3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human G2E3 (NP_0602392). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.