Learn More
R Langenbrinck™ g-TUBE™ for DNA Shearing
Product Code. 15240099
Encompass
Change view
Click to view available options
Quantity:
10 Individually Wrapped Tubes
Unit Size:
Each
Product Code. Quantity unitSize
15240099 10 Individually Wrapped Tubes Each
1 options
Product Code. 15240099 Supplier R Langenbrinck™ Supplier No. 520079
A single-use device that shears genomic DNA into selected fragment sizes ranging from 6kb to 20kb. The only equipment needed is a compatible bench-top centrifuge.
  • Uses centrifugal force to push the sample through a precisely manufactured orifice, producing the shearing forces in the sample that fragment DNA
  • Fragment size is selected by adjusting the centrifuge rotor speed which alters the flow rate through the orifice and thus the shearing force
  • Higher centrifugation RPM will produce shorter fragments and the protocol provided by Covaris contains the settings to generate fragment distributions centered on 6, 8, 10 or 20kbp
  • The entire process takes only 3 minutes and up to 24 samples can be processed simultaneously with a high reproducibility
Specifications
Buffer DI Water or TE or 10 mM Tris-Cl pH 8.5
Quantity 10 Individually Wrapped Tubes
Separation Range 6 kb to 20 kb