missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G Protein-Coupled Receptor Kinase 5 Rabbit Anti-Human,Mouse, Polyclonal, Abnova&Trade;
Rabbit Polyclonal Antibody
Brand: Abnova PAB4519.400uL
This item is not returnable.
View return policy
Description
G protein-coupled receptor kinase 5 Polyclonal antibody specifically detects G protein-coupled receptor kinase 5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
| G protein-coupled receptor kinase 5 | |
| Polyclonal | |
| ELISA (1:1000) Western Blot (1:50-100) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. |
|
| GPRK5 | |
| A synthetic peptide (conjugated with KLH) corresponding to 561-590 amino acids at C-terminus of human GRK5. | |
| RUO | |
| 2869 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| In PBS (0.09% sodium azide) | |
| Rabbit | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction