missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G Protein alpha z Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31604
This item is not returnable.
View return policy
Description
G Protein alpha z Polyclonal specifically detects G Protein alpha z in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| G Protein alpha z | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| GNAZ | |
| Recombinant protein encompassing a sequence within the center region of human GNAZ. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 2781 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| G(x) alpha chain, guanine nucleotide binding protein (G protein), alpha z polypeptide, guanine nucleotide-binding protein G(z) subunit alpha, gz-alpha, transducin alpha | |
| Rabbit | |
| 41 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction