missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ G protein alpha Inhibitor 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-30851-100ug

 View more versions of this product

Product Code. 30067012

  • £386.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

G protein alpha Inhibitor 2 Polyclonal antibody specifically detects G protein alpha Inhibitor 2 in Bovine,Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

G protein alpha Inhibitor 2
Polyclonal
Western Blot : 5-10 μg/mL
alpha-2 subunit, guanine nucleotide binding protein (G protein), alpha inhibiting activitypolypeptide 2
Rabbits were immunized with a peptide corresponding to amino acids 345-354 of G protein alpha Inhibitor 2 and 346-355 of Gi 2.
100 μg
Signal Transduction
2771
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Antigen Affinity-purified
RUO
Primary
Bovine, Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.