missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ G protein alpha Inhibitor 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30851-25ug
This item is not returnable.
View return policy
Description
G protein alpha Inhibitor 2 Polyclonal antibody specifically detects G protein alpha Inhibitor 2 in Bovine,Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| G protein alpha Inhibitor 2 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| alpha-2 subunit, guanine nucleotide binding protein (G protein), alpha inhibiting activitypolypeptide 2 | |
| Rabbits were immunized with a peptide corresponding to amino acids 345-354 of G protein alpha Inhibitor 2 and 346-355 of Gi 2. | |
| 25 μg | |
| Signal Transduction | |
| 2771 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction