missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G protein alpha Antibody (OTI12D9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-70837CL1
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
G protein alpha Monoclonal antibody specifically detects G protein alpha in Human samples. It is validated for Western Blot
Especificaciones
| G protein alpha | |
| Monoclonal | |
| CoraFluor 1 | |
| Adenylate cyclase-stimulating G alpha protein, AHO, Alternative gene product encoded by XL-exon, Extra large alphas protein, GNAS complex locus, GNAS1GSA, GNASXL, GPSAC20orf45, GSP, guanine nucleotide binding protein (G protein), alpha stimulating activitypolypeptide 1, guanine nucleotide regulatory protein, guanine nucleotide-binding protein G(s) subunit alpha isoforms XLas, MGC33735, NESP, NESP55, neuroendocrine secretory protein, PHP1A, PHP1B, PHP1C, POH, protein ALEX, SCG6, secretogranin VI, XLalphas | |
| Full length human recombinant protein of human GNAS (NP_000507) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 2778 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Western Blot | |
| OTI12D9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido