missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G protein alpha 16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03472-100UL
This item is not returnable.
View return policy
Description
G protein alpha 16 Polyclonal specifically detects G protein alpha 16 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| G protein alpha 16 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| G alpha-15, G alpha-16, GNA16G-protein subunit alpha-15, G-protein subunit alpha-16, guanine nucleotide binding protein (G protein), alpha 15 (Gq class), guanine nucleotide-binding protein subunit alpha-15, Guanine nucleotide-binding protein subunit alpha-16 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-374 of human G protein alpha 16 (NP_002059.3). | |
| 100 μL | |
| Signal Transduction | |
| 2769 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction