missing translation for 'onlineSavingsMsg'
Learn More

G protein alpha 16 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18307914 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18307914 20 μL 20µL
18373195 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18307914 Supplier Novus Biologicals Supplier No. NBP30324920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

G protein alpha 16 Polyclonal specifically detects G protein alpha 16 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen G protein alpha 16
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias G alpha-15, G alpha-16, GNA16G-protein subunit alpha-15, G-protein subunit alpha-16, guanine nucleotide binding protein (G protein), alpha 15 (Gq class), guanine nucleotide-binding protein subunit alpha-15, Guanine nucleotide-binding protein subunit alpha-16
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 112-374 of human G protein alpha 16 (NP_002059.3).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2769
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.