missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G protein alpha 16 Antibody (OTI6B3), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70836B
This item is not returnable.
View return policy
Description
G protein alpha 16 Monoclonal specifically detects G protein alpha 16 in Human, Rat samples. It is validated for Western Blot.
Specifications
| G protein alpha 16 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| GNA15 | |
| Recombinant protein fragment corresponding to amino acids 1-256 of human G protein alpha 16 (NP_002059) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| IgG1 |
| Western Blot | |
| OTI6B3 | |
| Western Blot | |
| G alpha-15, G alpha-16, GNA16G-protein subunit alpha-15, G-protein subunit alpha-16, guanine nucleotide binding protein (G protein), alpha 15 (Gq class), guanine nucleotide-binding protein subunit alpha-15, Guanine nucleotide-binding protein subunit alpha-16 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 2769.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction