missing translation for 'onlineSavingsMsg'
Learn More

G gamma14 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18371046 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371046 20 μL 20µL
18358774 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18371046 Supplier Novus Biologicals Supplier No. NBP30350220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

G gamma14 Polyclonal specifically detects G gamma14 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen G gamma14
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias G protein cone gamma 8 subunit, gamma-T2 subunit, G-GAMMA-8, G-gamma-9, G-GAMMA-C, GNG8, GNG9G gamma-C, GNGT8, guanine nucleotide binding protein (G protein), gamma transducing activitypolypeptide 2, guanine nucleotide binding protein gamma 9, Guanine nucleotide binding protein gamma transducing activity polypeptide 2, guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-T2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-69 of human G gamma14 (NP_113686.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2793
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.