missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-CSFR/CD114 Antibody (SR2290), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | G-CSFR/CD114 |
|---|---|
| Clone | SR2290 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18612124
|
Novus Biologicals
NBP3-21791-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671284
|
Novus Biologicals
NBP3-21791-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
G-CSFR/CD114 Monoclonal antibody specifically detects G-CSFR/CD114 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| G-CSFR/CD114 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1441 | |
| IgG | |
| Affinity purified |
| SR2290 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), G-CSF receptor, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor | |
| A synthesized peptide derived from human G-CSFR/CD114 (Uniprot #: Q99062) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title