missing translation for 'onlineSavingsMsg'
Learn More

G-CSFR/CD114 Antibody (723806) [Janelia Fluor™ 525], Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals FAB60391JF525

Product Code. 30158756

  • £459.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

G-CSFR/CD114 Monoclonal antibody specifically detects G-CSFR/CD114 in Mouse samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

G-CSFR/CD114
Monoclonal
Janelia Fluor 525
CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), G-CSF receptor, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor
Mouse myeloma cell line NS0-derived recombinant mouse G-CSF R/CD114, Cys26-Asp626, Accession # P40223
RUO
1441
Store at 4°C in the dark.
IgG2a
Flow Cytometry, CyTOF
723806
50mM Sodium Borate
Rat
0.1 mL
Primary
Mouse
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.