missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-CSF Rabbit anti-Human, Macaque, Clone: CSF3/3166R, Novus Biologicals™
Rabbit Monoclonal Antibody
£431.00
Specifications
| Antigen | G-CSF |
|---|---|
| Clone | CSF3/3166R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
G-CSF Monoclonal specifically detects G-CSF in Human, Rhesus Macaque samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| G-CSF | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research | |
| 10 mM PBS with 0.05% BSA | |
| 1440 | |
| IgG | |
| Protein A or G purified |
| CSF3/3166R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Macaque | |
| C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
| Recombinant full-length human G-CSF protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title