missing translation for 'onlineSavingsMsg'
Learn More

PeproTech G-CSF Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

£177.00 - £1468.00

Specifications

Antigen G-CSF
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17865763
View Documents
PeproTech
500-P43-50UG
50 μg
£177.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17895643
GREENER_CHOICE
View Documents
PeproTech
500-P43-100UG
100 μg
£228.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17835053
GREENER_CHOICE
View Documents
PeproTech
500-P43-1MG
1 mg
£1468.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AA Sequence of recombinant protein: TPLGPASSLP QSFLLKCLEQ VRKIQGDGAA LQEKLCATYK LCHPEELVLL GHSLGIPWAP LSSCPSQALQ LAGCLSQLHS GLFLYQGLLQ ALEGISPELG PTLDTLQLDV ADFATTIWQQ MEELGMAPAL QPTQGAMPAF ASAFQRRAGG VLVASHLQSF LEVSYRVLRH LAQP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human G-CSF. Anti-Human G-CSF-specific antibody was purified by affinity chromatography employing an immobilized Human G-CSF matrix. Sandwich ELISA: To detect Human G-CSF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human G-CSF (500-P43Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human G-CSF. Western Blot: To detect hG-CSF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hG-CSF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

G-CSF (Granulocyte colony stimulating factor) is a naturally occurring cytokine that stimulates the production and antibacterial function of neutrophils and monocytes. Human G-CSF is an 18.8 kDa protein containing 175 amino acid residues, and a soluble isoform of the G-CSF receptor has been described. The pleotropic cytokine is produced by activated monocytes, macrophages, endothelial cells, fibroblasts, astrocytes, osteoblasts and bone marrow cells. G-CSF has been shown to have specific effects on the proliferation, differentiation and activation of hematopoietic cells. G-CSF is also expressed by various transformed cells such as carcinoma cells and myeloblastic leukemia cells. G-CSF is encoded by two distinct DNA sequences, resulting in a full size, high activity and a shorter, low activity isoform of G-CSF. G-CSF is highly conserved among species and has been shown to exert its biological functions through interaction with its receptor expressed on the surface of hematopoietic progenitors, neutrophilic granulocytes and certain carcinoma cell lines. Clinical use of G-CSF has been approved for several therapeutic applications, treatment of neonatal infections, therapy of acute myocardial infarction, granulocyte transfusion in patients with neutropenia, in severe infections and sepsis, therapy in chronic autoimmune neutropenia, treatment of acute myeloid leukemias, Sweet's syndrome and AIDS. Further, G-CSF has been shown to be a marker protein for different carcinomas such as bladder cancer and dysfunction of the protein has been linked to Kostmann Syndrome.
Specifications

Specifications

G-CSF
ELISA, Western Blot
Unconjugated
Rabbit
Human
P09919
1440
E.coli-derived Recombinant Human G-CSF.
Antigen affinity chromatography
Csf3
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF 3; CSF3; CSF3OS; Csfg; filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; granulocyte-colony stimulating factor; gsf3; lenograstim; MGC45931; MGI-IG; Pluripoietin; RP23-395E10.4
Csf3
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.