Learn More
DiscoverX G-CSF (Human)
Product Code. 17362469
Encompass
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. 17362469
Brand: DiscoverX 921143
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
G-CSF (Human) is an agonist for CSF1R, CSF2RA, and CSF3R. Provided as 10μg of dry powder alongside reconstitution buffer.
Granulocyte-colony stimulating factor (G-CSF) is a glycoprotein growth factor, cytokine, and hormone that stimulates the production of granulocytes and stem cells in the bone marrow. Also functions in the differentiation, proliferation, and survival of neutrophils via the JAK/STAT, Ras/MAPK, and PI3K/Akt signal transduction pathways. Acts as an agonist for CSF1R, CSF2RA, and CSF3R. Provided as 10μg of dry powder alongside reconstitution buffer.