missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-CSF Antibody (CSF3/3166R), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08381FR
This item is not returnable.
View return policy
Description
G-CSF Monoclonal antibody specifically detects G-CSF in Human, Macaque samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| G-CSF | |
| Monoclonal | |
| DyLight 680 | |
| C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
| Recombinant full-length human G-CSF protein (Uniprot: P09919) | |
| 100 μg | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research | |
| 1440 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CSF3/3166R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction