missing translation for 'onlineSavingsMsg'
Learn More

G-749, MedChemExpress

Product Code. 16431337
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16431337

Brand: Medchem Express HY12333

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress G-749 is a potent, oral active and ATP competitive FLT3 inhibitor, with IC50s of 0.4 nM and 0.6 nM for FLT3 wild type and FLT3-D835Y, respectively. G-749 can be used for the research of drug resistance for acute myeloid leukemia (AML).

G-749 is a potent, oral active and ATP competitive FLT3 inhibitor, with IC50s of 0.4 nM and 0.6 nM for FLT3 wild type and FLT3-D835Y, respectively. G-749 can be used for the research of drug resistance for acute myeloid leukemia (AML).

Specifications

Chemical Name or Material G-749
CAS 1457983-28-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H25BrN6O2
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (47.95 mM; Need ultrasonic)
SMILES O=C1C2=C(N=C(NC3CCN(C)CC3)N=C2NC4=CC=C(OC5=CC=CC=C5)C=C4)C(Br)=CN1
Molecular Weight (g/mol) 521.41
Formula Weight 521.41
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.