missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-5555, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
Description
G-5555 is a potent p21-activated kinase 1 (PAK1) inhibitor with Kis of 3.7 nM and 11 nM for PAK1 and PAK2, respectively.
Specifications
Specifications
| Chemical Name or Material | G-5555 |
| CAS | 1648863-90-4 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C25H25ClN6O3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 25 mg/mL (50.71 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | ClC(C=C(C1=NC(C)=CC=C1)C=C2)=C2C3=CC4=CN=C(NC)N=C4N(C[C@@H]5OC[C@@H](N)CO5)C3=O |
| Molecular Weight (g/mol) | 492.96 |
| Formula Weight | 492.96 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction