missing translation for 'onlineSavingsMsg'
Learn More

G-5555 hydrochloride, MedChemExpress

Product Code. 16497828
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497828 10 mg 10mg
16483666 25 mg 25mg
16440298 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497828 Supplier Medchem Express Supplier No. HY19635A
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress G-5555 hydrochloride is a potent and selective p21-activated kinase 1 (PAK1) inhibitor with a Ki of 3.7 nM.

G-5555 hydrochloride is a potent and selective p21-activated kinase 1 (PAK1) inhibitor with a Ki of 3.7 nM.

Specifications

Chemical Name or Material G-5555 hydrochloride
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H26Cl2N6O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (188.89 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (31.49 mM; Need ultrasonic)
SMILES ClC(C=C(C1=NC(C)=CC=C1)C=C2)=C2C3=CC4=CN=C(NC)N=C4N(C[C@@H]5OC[C@@H](N)CO5)C3=O.[H]Cl
Molecular Weight (g/mol) 529.42
Formula Weight 529.42
Percent Purity 96.76%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.