missing translation for 'onlineSavingsMsg'
Learn More

G-5555 hydrochloride, MedChemExpress

Product Code. 16483666
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16483666

Brand: Medchem Express HY19635A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress G-5555 hydrochloride is a potent and selective p21-activated kinase 1 (PAK1) inhibitor with a Ki of 3.7 nM.

G-5555 hydrochloride is a potent and selective p21-activated kinase 1 (PAK1) inhibitor with a Ki of 3.7 nM.

Specifications

Chemical Name or Material G-5555 hydrochloride
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H26Cl2N6O3
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (188.89 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (31.49 mM; Need ultrasonic)
SMILES ClC(C=C(C1=NC(C)=CC=C1)C=C2)=C2C3=CC4=CN=C(NC)N=C4N(C[C@@H]5OC[C@@H](N)CO5)C3=O.[H]Cl
Molecular Weight (g/mol) 529.42
Formula Weight 529.42
Percent Purity 96.76%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.