missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ G 5555

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
G 5555 is a high affinity group 1 PAK (pan-PAK1, 2 and 3) inhibitor (Ki = 3.7 nM; pMEK IC50 = 69 nM). Of 235 kinases tested, other than PAK1, eight (PAK2, PAK3, KHS1, Lck, MST3, MST4, SIK2, and YSK1) were inhibited >70%. Exhibits low hERG channel activity (<50% inhibition at 10 μM). Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 8-[(trans-5-Amino-1,3-dioxan-2-yl)methyl]-6-[2-chloro-4-(6-methyl-2-pyridinyl)phenyl]-2-(methylamino)-pyrido[2,3-d]pyrimidin-7(8H)-one |
| CAS | 1648863-90-4 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Molecular Formula | C25H25ClN6O3 |
| Formula Weight | Observed MW: 492.96 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction