missing translation for 'onlineSavingsMsg'
Learn More

Melford G-418 Disulfate Solution

Product Code. 30018322
Change view
Click to view available options
Quantity:
10 mL
20 mL
50 mL
Unit Size:
10mL
20mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018322 50 mL 50mL
30018321 20 mL 20mL
30018320 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018322 Supplier Melford Supplier No. G6450050.0
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

G-418 Disulfate, Solution is a widely used antibiotic in the life science marketplace.

G-418 Disulfate, Solution is a widely used antibiotic in the life science marketplace. It is commonly employed as a selective agent in cell culture and molecular biology experiments. G-418, also known as Geneticin, is a member of the aminoglycoside family of antibiotics.

In the life science field, G-418 Disulfate Solution is primarily utilized for the selection and maintenance of cells that have been genetically modified to express the neomycin resistance gene. This resistance gene allows the cells to survive in the presence of G-418, while non-modified cells are unable to survive.

Researchers and scientists working in areas such as molecular biology, genetic engineering, and biotechnology rely on G-418 Disulfate Solution for various applications. It is frequently used as a selectable marker in plasmids and other vectors to ensure that only cells carrying the desired genetic material survive and proliferate.

G-418 Disulfate Solution is compatible with a wide range of mammalian, insect, and plant cell lines, making it versatile for different cell culture systems. It is typically added to the culture medium at specific concentrations to inhibit the growth of non-resistant cells while allowing the growth of genetically modified cells.

Moreover, G-418 Disulfate Solution can be used to generate stable cell lines expressing recombinant proteins or studying gene function. Its broad-spectrum activity and compatibility with different cell types make it an important tool for researchers studying molecular mechanisms, protein expression, and genetic manipulation.

Specifications

Chemical Name or Material G-418 Disulfate, Solution
CAS 108321-42-2
Quantity 50 mL
Molecular Formula C20H40N4O10-2H2SO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.