missing translation for 'onlineSavingsMsg'
Learn More

G-418 disulfate, MedChemExpress

Product Code. p-200096823
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16421699

Brand: Medchem Express HY17561

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress G-418 disulfate (Geneticin sulfate), is an aminoglycoside antibiotic, inhibits protein synthesis in eukaryotes and prokaryotes. G-418 disulfate is commonly used as a selective agent for eukaryotic cells.

G-418 disulfate (Geneticin sulfate), is an aminoglycoside antibiotic, inhibits protein synthesis in eukaryotes and prokaryotes. G-418 disulfate is commonly used as a selective agent for eukaryotic cells.

Specifications

Chemical Name or Material G-418 disulfate
CAS 108321-42-2
Color White
Purity Grade Notes Research
Molecular Formula C20H44N4O18S2
Quantity 1 g
Synonym Geneticin sulfate Antibiotic G-418 sulfate
Solubility Information H2O : 125 mg/mL (180.45 mM; Need ultrasonic)
SMILES O=S(O)(O)=O.O[C@@H]([C@@H]1O[C@@]([C@@H]([C@@H](O)[C@@H]2O)N)([H])O[C@]2([H])[C@H](O)C)[C@H]([C@@H](C[C@@H]1N)N)O[C@](OC[C@](C)(O)[C@@H]3NC)([H])[C@@H]3O.O=S(O)(O)=O
Molecular Weight (g/mol) 692.71
Formula Weight 692.71
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.