missing translation for 'onlineSavingsMsg'
Learn More

Melford G-418 Disulfate

Product Code. 30018319
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018319 5 g 5g
30018317 1 g 1g
30018318 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018319 Supplier Melford Supplier No. G640005.0
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

G-418 Disulfate, also known as Geneticin, is a widely used antibiotic in the life science marketplace.

G-418 Disulfate, also known as Geneticin, is a widely used antibiotic in the life science marketplace. It is commonly utilized as a selective agent in cell culture systems and molecular biology applications.

G-418 is derived from the bacterium Micromonospora rhodorangea and belongs to the class of aminoglycoside antibiotics. It acts by inhibiting protein synthesis in cells, specifically targeting the 80S ribosome and interfering with translocation.

In the life science marketplace, G-418 Disulfate finds extensive use as a selective agent for cells that have been genetically modified or transfected with a G-418 resistance gene. This resistance gene is often incorporated into plasmid vectors or recombinant DNA constructs to allow for the selection and maintenance of cells that have successfully taken up the desired genetic material.

The selective nature of G-418 Disulfate makes it an invaluable tool in molecular biology research. By incorporating the G-418 resistance gene and applying G-418 treatment, researchers can specifically select cells that have successfully integrated the desired genetic material.

This antibiotic selection system is commonly used in techniques such as stable cell line generation, gene expression studies, and protein production.

Specifications

Chemical Name or Material G-418 Disulfate
CAS 108321-42-2
Quantity 5 g
Molecular Formula C20H40N4O10-2H2SO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.