missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FZR1, Mouse anti-Human, Clone: 3E12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00051343-M09.100ug
This item is not returnable.
View return policy
Description
Sequence: MDQDYERRLLRQIVIQNENTMPRVTEMRRTLTPASSPVSSPSKHGDRFIPSRAGANWSVNFHRINENEKSPSQNRKAKDATSDNGKDGLAYSALLKNELLGSpecifications
| FZR1 | |
| Monoclonal | |
| Unconjugated | |
| fizzy/cell division cycle 20 related 1 (Drosophila) | |
| CDC20C/CDH1/FZR/FZR2/HCDH/HCDH1/KIAA1242 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3E12 | |
| In 1x PBS, pH 7.4 | |
| NM_016263 | |
| FZR1 | |
| FZR1 (NP_057347.2, 1 a.a. ∼ 101 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51343 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction