missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FZR1/CDH1 Monoclonal antibody specifically detects FZR1/CDH1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 610 SE |
| Gene Alias | CDC20C, CDH1, Cdh1/Hct1 homolog, CDH1CDC20-like 1b, fizzy/cell division cycle 20 related 1 (Drosophila), Fzr, FZR2, FZRCDC20-like protein 1, hCDH1, HCDHFYR, KIAA1242fizzy-related protein homolog |
| Immunogen | His-tagged recombinant FZR1/CDH1 |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?