missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FZD9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115259
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FZD9.
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney.
Specifications
| FZD9 | |
| Polyclonal | |
| Unconjugated | |
| FZD9 | |
| CD349; CD349 antigen; Frizzled 3; frizzled 9; frizzled 9, seven transmembrane spanning receptor; frizzled class receptor 9; frizzled family receptor 9; frizzled homolog 9; frizzled homolog 9 (Drosophila); Frizzled3; Frizzled-3; Frizzled9; frizzled-9; Frizzled-like protein 9; fz-9; FZD3; FZD9; FzE6; hFz9; mFz3; mfz9; rFz9 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14371, 8326 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O00144, Q9R216 | |
| FZD9 | |
| A synthesized peptide derived from human FZD9(Accession O00144), corresponding to amino acid residues R556-L591. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction