Learn More
Invitrogen™ FZD9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597000
Description
Immunogen sequence: ERLNMDFWRL RATEQPCAAA AGPGGRRDCS LPGGSVPTVA VFMLKIFMSL VVGITSGVWV WSSKTFQTWQ SLCYRKIAAG RARAKACRAP GSYGRGTHCH YKAPTVVLHM TKTDPSLENP THL; Positive Samples: TE-1, 293T, HepG2, THP-1, U-251MG, Mouse heart, Mouse brain, Rat skeletal muscle; Cellular Location: Cell membrane, Multi-pass membrane protein.
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney.
Specifications
| FZD9 | |
| Polyclonal | |
| Unconjugated | |
| FZD9 | |
| CD349; CD349 antigen; Frizzled 3; frizzled 9; frizzled 9, seven transmembrane spanning receptor; frizzled class receptor 9; frizzled family receptor 9; frizzled homolog 9; frizzled homolog 9 (Drosophila); Frizzled3; Frizzled-3; Frizzled9; frizzled-9; Frizzled-like protein 9; fz-9; FZD3; FZD9; FzE6; hFz9; mFz3; mfz9; rFz9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14371, 266608, 8326 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.64 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00144, Q8K4C8, Q9R216 | |
| FZD9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 469-591 of human FZD9 (NP_0034991). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.