missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FZD8 Recombinant Rabbit Monoclonal Antibody (23GB5470), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | FZD8 |
|---|---|
| Clone | 23GB5470 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088403
![]() |
Invitrogen™
MA552860 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089362
|
Invitrogen™
MA552861 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FZD8 Recombinant Monoclonal antibody specifically detects FZD8 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| FZD8 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9H461 | |
| 8325 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5470 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| frizzled 8, seven transmembrane spanning receptor; frizzled class receptor 8; frizzled family receptor 8; frizzled homolog 8; frizzled homolog 8 (Drosophila); frizzled-8; Fz8; Fz-8; Fzd8; hFZ8; HGNC:4046 | |
| A synthesized peptide derived from human Frizzled 8 (640-694AA). | |
| Primary | |
| -20°C | |
| Fzd8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title