Learn More
Invitrogen™ FZD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592165
Description
Immunogen sequence: MRDPGAAAPL SSLGLCALVL ALLGALSAGA GAQPYHGEKG ISVPDHGFCQ PISIPLCTDI AYNQTILPNL LGHTNQEDAG LEVHQFYPLV KVQCSPELRF FLCSMYAPVC TVLDQAIPPC RSLCERARQG CEALMNKFGF QWPERLRCEN FPVHGAGEIC VGQNTSDGSG GPGGGPTAYP TAPYLPDLPF TALPPGASDG RGRPAFPFSC PRQLKVPPYL GYRFLGERDC GAPCEPGRAN GLMYFKEEER RFARLWVGVW; Positive Samples: SW480, Mouse kidney; Cellular Location: Cell membrane, Membrane, Multi-pass membrane protein.
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD7 protein contains an N-terminal signal sequence, 10 cysteine residues typical of the cysteine-rich extracellular domain of Fz family members, 7 putative transmembrane domains, and an intracellular C-terminal tail with a PDZ domain-binding motif. FZD7 gene expression may downregulate APC function and enhance beta-catenin-mediated signals in poorly differentiated human esophageal carcinomas.
Specifications
| FZD7 | |
| Polyclonal | |
| Unconjugated | |
| FZD7 | |
| frizzled 7, seven transmembrane spanning receptor; frizzled class receptor 7; frizzled family receptor 7; frizzled homolog 7; frizzled homolog 7 (Drosophila); Frizzled, drosophila, homolog of, 7; frizzled-7; Fz7; Fz-7; FZD7; FzE3; hFz7; LOC100360552; mFz7; rCG22430-like; rizzled 7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14369, 8324 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75084, Q61090 | |
| FZD7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human FZD7 (NP_0034981). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.