missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FZD7 Monoclonal Antibody (151143), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524378
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant mouse Frizzled-1, -3, -4 or -8 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
FZD7 Monoclonal antibody specifically detects FZD7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
FZD7 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O75084, Q61090 | |
FZD7 | |
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-7 Gln33-Leu185 | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry | |
151143 | |
Unconjugated | |
FZD7 | |
frizzled 7, seven transmembrane spanning receptor; frizzled class receptor 7; frizzled family receptor 7; frizzled homolog 7; frizzled homolog 7 (Drosophila); Frizzled, drosophila, homolog of, 7; frizzled-7; Fz7; Fz-7; FZD7; FzE3; hFz7; LOC100360552; mFz7; rCG22430-like; rizzled 7 | |
Rat | |
Protein A/G | |
RUO | |
14369, 8324 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |