missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FZD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575512
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The exact function of the protein encoded by this gene is not known. However, this gene product contains a KRAB domain (which is involved in protein-protein interactions) at the N-terminus, and a transposase domain at the C-terminus, suggesting that it may belong to the family of DNA-mediated transposons in human.
Specifications
| FZD6 | |
| Polyclonal | |
| Unconjugated | |
| FZD6 | |
| frizzled 6; frizzled 6, seven transmembrane spanning receptor; frizzled class receptor 6; frizzled family receptor 6; frizzled homolog 6; frizzled homolog 6 (Drosophila); Frizzled6; frizzled-6; FZ6; Fz-6; Fzd6; FZD-6; HFZ6; mFz6; NDNC10; seven transmembrane helix receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14368, 282581, 8323 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60353, Q61089 | |
| FZD6 | |
| Synthetic peptide corresponding to amino acids 167-212 of Human FZD6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction