missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FZD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533546
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset (100%).
The exact function of the protein encoded by this gene is not known. However, this gene product contains a KRAB domain (which is involved in protein-protein interactions) at the N-terminus, and a transposase domain at the C-terminus, suggesting that it may belong to the family of DNA-mediated transposons in human.
Specifications
| FZD6 | |
| Polyclonal | |
| Unconjugated | |
| FZD6 | |
| frizzled 6; frizzled 6, seven transmembrane spanning receptor; frizzled class receptor 6; frizzled family receptor 6; frizzled homolog 6; frizzled homolog 6 (Drosophila); Frizzled6; frizzled-6; FZ6; Fz-6; Fzd6; FZD-6; HFZ6; mFz6; NDNC10; seven transmembrane helix receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8323 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O60353 | |
| FZD6 | |
| Synthetic 16 amino acid peptide from N-terminal extracellular domain of human Frizzled-6. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction