Learn More
Invitrogen™ FZD6 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547151
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant human Frizzled-6 is observed, and less than 1% cross-reactivity with recombinant mouse (rm) Frizzled-1, rmFrizzled-2, rmFrizzled-3, rmFrizzled-4, rmFrizzled-7, rmFrizzled-8, and rmFrizzled-9 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The exact function of the protein encoded by this gene is not known. However, this gene product contains a KRAB domain (which is involved in protein-protein interactions) at the N-terminus, and a transposase domain at the C-terminus, suggesting that it may belong to the family of DNA-mediated transposons in human.
Specifications
| FZD6 | |
| Polyclonal | |
| Unconjugated | |
| FZD6 | |
| frizzled 6; frizzled 6, seven transmembrane spanning receptor; frizzled class receptor 6; frizzled family receptor 6; frizzled homolog 6; frizzled homolog 6 (Drosophila); Frizzled6; frizzled-6; FZ6; Fz-6; Fzd6; FZD-6; HFZ6; mFz6; NDNC10; seven transmembrane helix receptor | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 14368 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q61089 | |
| FZD6 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-6 His19-Val153. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.