missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fyn Monoclonal Antibody (1S), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112607
This item is not returnable.
View return policy
Description
This antibody recognizes epitope aa 26-75. Staining of formalin-fixed paraffin embedded tissue sections requires boiling the sections in 10 mM citrate buffer, pH 6.0, for 10-20 minutes followed by cooling at room temperature for 20 minutes.
FYN is a SRC-related kinase expressed in brain, skeletal muscle and adipose tissues. Fyn functions in a large number of pathways and has been shown to play a role in memory. FYN is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.
Specifications
| Fyn | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P06241, P39688, Q62844 | |
| Fyn | |
| Recombinant protein corresponding to amino acid residues 1-206 of murine p59fynprotein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1S | |
| Unconjugated | |
| Fyn | |
| AI448320; AW552119; C syn protooncogene; c-syn protooncogene; EC 2.7.10.2; Fyn; fyn {ECO:0000312; FYN oncogene related to SRC, FGR, YES; Fyn proto-oncogene; FYN proto-oncogene, Src family tyrosine kinase; kinase Fyn; MGC45350; OKT3-induced calcium influx regulator; OTTHUMP00000017916; OTTHUMP00000017917; P59 p59-Fyn; p59-Fyn; Proto-oncogene c-Fyn; Protooncogene Syn; proto-oncogene Syn; proto-oncogene tyrosine-protein kinase Fyn; RGD:2641}; RP1-66H14.1; SLK; Src Kinase p59; Src like kinase; src/yes-related novel; src-like kinase; SYN; tyrosine kinase p59fyn(T); tyrosine-protein kinase Fyn | |
| Mouse | |
| Protein G | |
| RUO | |
| 14360, 25150, 2534 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction