missing translation for 'onlineSavingsMsg'
Learn More

Fyn Antibody (361228) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30731667 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731667 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731667 Supplier Novus Biologicals Supplier No. FAB3574MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fyn Monoclonal antibody specifically detects Fyn in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fyn
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 361228
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10, EC 2.7.10.2, FYN oncogene related to SRC, FGR, YES, MGC45350, OKT3-induced calcium influx regulator, p59-Fyn, protein-tyrosine kinase fyn, Proto-oncogene c-Fyn, Proto-oncogene Syn, proto-oncogene tyrosine-protein kinase fyn, SLKc-syn protooncogene, src/yes-related novel, Src-like kinase, SYN, tyrosine kinase p59fyn(T), tyrosine-protein kinase Fyn
Host Species Mouse
Immunogen E. coli-derived recombinant human Fyn, Met1-Gly83, Accession # P06241
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2534
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.