missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FYN A, GST Tag Recombinant Protein

Product Code. 15448983
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 15448983

Brand: Invitrogen™ PV6348

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Full-length recombinant human FYN A was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Validated in LanthaScreen™ Kinase Binding Assay Protocols available for simple use and optimized performance Multiple pack sizes available for convenient use Related Product:LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant The gene accession number for FYN A is NP_002028.1.

Fyn A (Fyn isoform A) is a member of the SRC tyrosine kinase oncogene family showing high homology to YES1, FGR and SRC. Fyn has been shown to phosphorylate Dab1, an intracellular adaptor protein that interacts with amyloid precursor protein (APP) and apoE receptor 2 (apoEr2). The interaction of Fyn and Dab1 regulates the phosphorylation, trafficking, and processing of APP and apoEr2. Fyn expression has been shown to be significantly increased in Chronic Myelogenous Leukemia (CML). Knockdown of Fyn with shRNA slows leukemia cell growth, inhibits clonogenicity, and leads to increased sensitivity to imatinib.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P06241
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.1mM PMSF, 150mM NaCl, 25% glycerol, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 2534
Molecular Weight (g/mol) 87.1 kDa
Name Human FYN A, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias FYN
Common Name FYN Isoform A
Gene Symbol FYN
Protein Length 1-537
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-537
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Cytoplasmic Tyrosine Kinase
Expression System Baculovirus
Protein Form Recombinant
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.