missing translation for 'onlineSavingsMsg'
Learn More

FYCO1 Antibody (OTI9D1) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30688093 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688093 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688093 Supplier Novus Biologicals Supplier No. NBP345026JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FYCO1 Monoclonal antibody specifically detects FYCO1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FYCO1
Applications Western Blot
Classification Monoclonal
Clone OTI9D1
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias DKFZp779K1152, FLJ13335, FYVE and coiled-coil domain containing 1, FYVE and coiled-coil domain-containing protein 1, MGC126517, RUFY3, RUN and FYVE domain containing 3, ZFYVE7MGC126519, Zinc finger FYVE domain-containing protein 7
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1102-1187 of human FYCO1 (NP_078789) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79443
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.