missing translation for 'onlineSavingsMsg'
Learn More

FYCO1 Antibody (OTI8G7), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-45030-30ul

 View more versions of this product

Product Code. 30306129

  • £195.00 / 30µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FYCO1 Monoclonal antibody specifically detects FYCO1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FYCO1
Monoclonal
Unconjugated
PBS (pH 7.3) 1% BSA, 50% glycerol
Mouse
>95%
RUO
79443
Store at -20 degrees C. Avoid freeze/thaw cycles.
Purified
Western Blot
OTI8G7
Western Blot 1:2000
DKFZp779K1152, FLJ13335, FYVE and coiled-coil domain containing 1, FYVE and coiled-coil domain-containing protein 1, MGC126517, RUFY3, RUN and FYVE domain containing 3, ZFYVE7MGC126519, Zinc finger FYVE domain-containing protein 7
Human recombinant protein fragment corresponding to amino acids 1102-1187 of human FYCO1(NP_078789) produced in E.coli.
30 μL
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.