missing translation for 'onlineSavingsMsg'
Learn More

FYB/ADAP/SLAP130 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18416043 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416043 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416043 Supplier Novus Biologicals Supplier No. NBP176811UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FYB/ADAP/SLAP130 Polyclonal antibody specifically detects FYB/ADAP/SLAP130 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FYB/ADAP/SLAP130
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ADAP, Adhesion and degranulation promoting adaptor protein, adhesion and degranulation-promoting adaptor protein, FYB-120/130SLAP-130FYN-binding protein (FYB-120/130), FYN binding protein, FYN-binding protein, FYN-T-binding protein, p120/p130, PRO0823, SLAP130, SLP-76-associated phosphoprotein
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human ADAP. The immunogen is located within amino acids 60 - 110 of ADAP. Amino Acid Squence: SEEKPDKEPKPPFLKPT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2533
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.