missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FXYD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572700
This item is not returnable.
View return policy
Description
FXYD7 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The specific function of this protein remains unknown.This reference sequence was derived from multiple replicate ESTs and validated by similar human genomic sequence. This gene encodes a member of a family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The approved human gene nomenclature for the family is FXYD-domain containing ion transport regulator. Transmembrane topology has been established for two family members (FXYD1 and FXYD2), with the N-terminus extracellular and the C-terminus on the cytoplasmic side of the membrane. FXYD2, also known as the gamma subunit of the Na, K-ATPase, regulates the properties of that enzyme. FXYD1 (phospholemman), FXYD2 (gamma), FXYD3 (MAT-8), FXYD4 (CHIF), and FXYD5 (RIC) have been shown to induce channel activity in experimental expression systems. This gene product, FXYD7, is novel and has not been characterized as a protein.
Specifications
| FXYD7 | |
| Polyclonal | |
| Unconjugated | |
| FXYD7 | |
| 1110035I01Rik; FLJ25096; FXYD domain containing ion transport regulator 7; FXYD domain-containing ion transport regulator 7; FXYD7; Unknown (protein for MGC:128487) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53822, 57780, 63848 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P58549, P59648, P59649 | |
| FXYD7 | |
| FXYD7 antibody for a 17 amino acid synthetic peptide near the amino terminus of human FXYD7. The immunogen is within the first 50 amino acids of FXYD7. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction