missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FXYD7 Polyclonal antibody specifically detects FXYD7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | FXYD7 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Gene Alias | FLJ25096, FXYD domain containing ion transport regulator 7, FXYD domain-containing ion transport regulator 7 |
| Host Species | Rabbit |
| Immunogen | FXYD7 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human FXYD7. The immunogen is located within the first 50 amino acids of FXYD7. Amino Acid Squence: ATPTQTPTKAPEEPDPF |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?