missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXYD6 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | FXYD6 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18335126
|
Novus Biologicals
NBP3-04818-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18334575
|
Novus Biologicals
NBP3-04818-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FXYD6 Polyclonal specifically detects FXYD6 in Mouse, Rat samples. It is validated for Western Blot.Specifications
| FXYD6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 53826 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| FXYD domain containing ion transport regulator 6, FXYD domain-containing ion transport regulator 6, phosphohippolin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-95 of human FXYD6 (NP_071286.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title