missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FXYD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565327
This item is not returnable.
View return policy
Description
Immunogen sequence: CKCSFNQKPRA PGDEEAQVEN LITANATEP Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
This gene encodes a member of the FXYD family of transmembrane proteins. This particular protein encodes phosphohippolin, which likely affects the activity of Na,K-ATPase. Multiple alternatively spliced transcript variants encoding the same protein have been described. Related pseudogenes have been identified on chromosomes 10 and X. Read-through transcripts have been observed between this locus and the downstream sodium/potassium-transporting ATPase subunit gamma (FXYD2, GeneID 486) locus.
Specifications
| FXYD6 | |
| Polyclonal | |
| Unconjugated | |
| FXYD6 | |
| 0610030I18Rik; FXYD domain containing ion transport regulator 6; FXYD domain-containing ion transport regulator 6; Fxyd6; Phosphohippolin; Php; PLM-like protein; Plp; UNQ521/PRO1056; Vascular endothelial cell-specific protein 6; VESP6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 53826 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H0Q3 | |
| FXYD6 | |
| Recombinant Human FXYD6. Recombinant protein control fragment (Product #RP-106023). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction