missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXYD5/Dysadherin Antibody (M031), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23350
This item is not returnable.
View return policy
Description
FXYD5/Dysadherin Monoclonal antibody specifically detects FXYD5/Dysadherin in Human samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications
| FXYD5/Dysadherin | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| M031 | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:50 | |
| DYSAD, dysadherin, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC | |
| Clone (M031) was generated from a proprietary antigen related to the native human FXYD5/Dysadherin protein expressed in MDA-MB-231 breast cancer cell line. [UniProt# Q96DB9] | |
| 0.1 mL | |
| Signal Transduction | |
| 53827 | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction