missing translation for 'onlineSavingsMsg'
Learn More

FXYD5/Dysadherin Antibody [DyLight 650], Novus Biologicals™

Product Code. 30737078 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737078 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737078 Supplier Novus Biologicals Supplier No. AF8300W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

FXYD5/Dysadherin Polyclonal antibody specifically detects FXYD5/Dysadherin in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen FXYD5/Dysadherin
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias DYSAD, dysadherin, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC
Host Species Goat
Immunogen E. coli-derived recombinant human FXYD5/Dysadherin, Gln22-Asp133, Accession # Q96DB9
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 53827
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.